HPLC Analysis of an Energy Drink on Ascentis® Express HILIC (ELSD Detection)
HPLC Analysis of an Energy Drink on Ascentis® Express HILIC (ELSD Detection)
Conditions
column Ascentis Express HILIC, 5 cm x 3 mm I.D., 2.7 µm particles (53967-U)
column temp. 35 °C
mobile phase [A] 100 mM ammonium acetate, pH 5 (titrated with acetic acid) [B] water [C] acetonitrile; (9:1:90, A:B:C:)
flow rate 0.6 mL/min
sample Energy Drink Diluted 1:9 in acetonitrile
injection 2 µL
detector ELSD, 55 °C, 3.5 bar nitrogen
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Sports and Energy Drinks
Featured Industry Food and Beverages