HPLC Analysis of an Energy Drink on Ascentis® Express RP-Amide (UV detection)
HPLC Analysis of an Energy Drink on Ascentis® Express RP-Amide (UV detection)
Conditions
column Ascentis Express RP-Amide, 5 cm x 3 mm I.D., 2.7 µm particles (53916-U)
column temp. 35 C
mobile phase [A] water with 0.1% TFA; [B] acetonitrile with 0.1% TFA
flow rate 0.6 ml/min
pressure 1146 psi (79 bar)
sample Energy Drink Diluted 1:9 in acetonitrile
injection 2 µl
detector UV, 254 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in caffeinated energy drinks.
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Sports and Energy Drinks
Featured Industry Food and Beverages