HPLC Analysis of an mPEG-peptide on Zenix® SEC Phases
HPLC Analysis of an mPEG-peptide on Zenix® SEC Phases
Conditions
column Zenix C SEC-300, 30 cm x 7.8mm I.D., 3 µm particles, 300 Ã… (Z777092)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles, 300 Ã… (Z777033)
column temp. 25 °C
mobile phase 150 mM Sodium Phosphate Buffer, pH 7.0
flow rate 1.0 mL/min
sample 20kD mPEG-MAL (methoxy-PEG-Maleimide) (1mg/mL)
injection 20 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies