HPLC Analysis of Anaesthetic Drugs on Ascentis® RP-Amide
HPLC Analysis of Anaesthetic Drugs on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate (pH 4.5 with acetic acid); [B] acetonitrile; (70:30, A:B)
flow rate 1 mL/min
sample 1 µg/mL each in mobile phase
injection 10 µL
detector ESI (+)
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anesthetic Drugs, Anesthetics