HPLC Analysis of Analgesics, Narcotics on Ascentis® Phenyl
HPLC Analysis of Analgesics, Narcotics on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate (pH 3.0 with formic acid); [B] acetonitrile; (90:10, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL in 90:10, water:methanol
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, Anesthetic Drugs, analgesics, Analgesics
Featured Industry Clinical
Pharmaceutical (small molecule)