Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Angiotensins on Discovery® BIO Wide Pore C18
HPLC Analysis of Angiotensins on Discovery® BIO Wide Pore C18
Conditions
column Discovery BIO Wide Pore C18, 15 cm x 4.6 mm I.D., 5 µm particles (568222-U)
column temp. ambient
mobile phase [A] (10 mM NH4OAc, pH 7):(50% CH3CN in 20 mM NH4OAc, pH 7), 65:35 ; [B] (10 mM NH4OAc, pH 7):(50% CH3CN in 20 mM NH4OAc, pH 7), 25:75
gradient 0-100% B in 12.5 min
flow rate 1 ml/min
injection 6 µl(10 mug each) in water
detector UV, 254 nm
Description
Categories Analytical Chromatography, Hormones
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC