HPLC Analysis of Angiotensins on Ascentis® RP-Amide
HPLC Analysis of Angiotensins on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 5 mM ammonium phosphate (pH 7.0 with ammonium hydroxide); [B] acetonitrile; (77.5:22.5, A:B)
flow rate 1.0 mL/min
sample as indicated in mobile phase
injection 10 µL
detector UV, 215 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Angiotensins, Hormones
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)