HPLC Analysis of Anions and Cations on SeQuant ZIC-HILIC
HPLC Analysis of Anions and Cations on SeQuant ZIC-HILIC
Conditions
column Sequant ZIC-HILIC, PEEK, 150 x 2.1 mm, 3.5 µm, 200 Ã… (1.50448.0001)
column temp. 40 C
mobile phase [A] acetonitrile; [B] 20 mM ammonium formate, pH 3
gradient 80% A held for 3 min; to 20% A in 7 min; held at 20% A for 3 min; to 80% A in 2 min; held at 80% A for 10 min
flow rate 0.3 ml/min
injection 2 µl
detector ELSD (SEDEX 85LT, 40C, 3.5 bar)
Description
Featured Industry Chemicals and Industrial Polymers