HPLC Analysis of Anthocyanins in Red Wine on Ascentis® C18
HPLC Analysis of Anthocyanins in Red Wine on Ascentis® C18
Conditions
column Ascentis C18, 25 cm x 4.6 mm I.D., 5 µm particles (581325-U)
column temp. 25 °C
mobile phase [A] water:formic acid (9:1), [B] acetonitrile:water:formic acid (5:4:1)
flow rate 1 mL/min
sample red wine diluted with water, centrifuged and passed through a 0.45 µm filter
injection 10 µL
detector VIS, 518 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Beer, Wine, and Spirits, Wine
Featured Industry Food and Beverages