HPLC Analysis of Anti-Inflammatory Drugs on Discovery® RP-Amide C16
HPLC Analysis of Anti-Inflammatory Drugs on Discovery® RP-Amide C16
Conditions
column Discovery RP-Amide C16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 30 °C
mobile phase (A) acetonitrile; (B) 25 mM KH2PO4, pH 3.0; (40:60, A:B)
flow rate 1 mL/min
injection 10 µL, 1 µg/mL each analyte
detector UV, 230 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anti-inflammatories
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC