HPLC Analysis of Anti-Inflammatory Drugs on Ascentis® C18
HPLC Analysis of Anti-Inflammatory Drugs on Ascentis® C18
Conditions
column Ascentis C8, 15 cm x 4.6 mm I.D., 5 µm particles (581424-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate (pH 3.0 with formic acid); [B] acetonitrile; (45:55, A:B)
flow rate 1.0 mL/min
sample as indicated in mobile phase
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anti-inflammatories