HPLC Analysis of Antianxiety Drugs on Ascentis® Express C18 vs. Standard C18, Demonstrating High Throughput on Ascentis Express
HPLC Analysis of Antianxiety Drugs on Ascentis® Express C18 vs. Standard C18, Demonstrating High Throughput on Ascentis Express
Conditions
column C18, 25 cm x 4.6 mm I.D., 5 µm particles and Ascentis Express C18, 10 cm x 4.6 mm I.D., 2.7 µm particles (53827-U)
column temp. ambient
mobile phase [A] water: [B] acetonitrile; (65:35, A:B)
flow rate 1 mL/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure, Anxiolytics, Sedatives