HPLC Analysis of Antibacterials/Antifungals on Ascentis® RP-Amide (Formate Mobile Phase)
HPLC Analysis of Antibacterials/Antifungals on Ascentis® RP-Amide (Formate Mobile Phase)
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate (pH 3.0 with formic acid); [B] acetonitrile; (75:25, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL each in mobile phase
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Clinical, antifungals, Antibacterials/Antifungals
Featured Industry Clinical
Pharmaceutical (small molecule)