HPLC Analysis of Antibiotic Sulfa Drugs on Ascentis® C18
HPLC Analysis of Antibiotic Sulfa Drugs on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 C
mobile phase [A] water with 1% acetic acid; [B] methanol; (85:15, A:B)
flow rate 1 ml/min
sample 50 µg/mL each in mobile phase
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics, Antimicrobials, Antibiotics