HPLC Analysis of Antibiotic Sulfa Drugs on Ascentis® RP-Amide
HPLC Analysis of Antibiotic Sulfa Drugs on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 1% acetic acid in water; [B] methanol, (85:15, A:B)
flow rate 1.0 mL/min
sample as indicated in 1% acetic acid in water
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics, Antimicrobials, Antibiotics