HPLC Analysis of Antibiotics on Discovery® C18
HPLC Analysis of Antibiotics on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 35 C
mobile phase [A] 25 mM potassium phosphate (pH 3.0); [B] acetonitrile
flow rate 1 ml/min
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC