HPLC Analysis of Anticonvulsants on Discovery® C18
HPLC Analysis of Anticonvulsants on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 20 °C
mobile phase [A] water: [B] acetonitrile; (70:30, A:B)
flow rate 2 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC