HPLC Analysis of Antihypertensive ACE Inhibitors on Discovery® RP-AmideC16
HPLC Analysis of Antihypertensive ACE Inhibitors on Discovery® RP-AmideC16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 35 C
mobile phase [A] acetonitrile; [B] 25 mM potassium phosphate, pH 2.3 (33:67, A:B)
flow rate 0.6 ml/min
injection 3 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, antihypertensives, Antihypertensives
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC