HPLC Analysis of Antihypertensive and Diuretic Drugs on Discovery® RP-Amide C16
HPLC Analysis of Antihypertensive and Diuretic Drugs on Discovery® RP-Amide C16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 30 C
mobile phase [A] methanol; [B] 25 mM potassium phosphate, dibasic, pH 7.0 (22:78, A:B)
flow rate 1 ml/min
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Clinical, Antihypertensives, antihypertensives
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC