HPLC Analysis of Antihypertensive Drugs on Ascentis® C18
HPLC Analysis of Antihypertensive Drugs on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 °C
mobile phase [A] 10 mM monobasic potassium phosphate (pH 7.0 with potassium hydroxide); [B]:acetonitrile
flow rate 1 mL/min
sample 50 µg/mL each in mobile phase
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, antihypertensives, Antihypertensives
Featured Industry Clinical