HPLC Analysis of Antioxidants on SUPELCOSIL™ LC-18
HPLC Analysis of Antioxidants on SUPELCOSIL™ LC-18
Conditions
column SUPELCOSIL LC-18, 15 cm x 4.6 mm I.D., 5 µm particles (58230-U)
mobile phase [A] 5% acetic acid in deionized water; [B] acetonitrile:methanol (1:1) 70% A/30% B to 100% B, linear gradient over 10 min, hold 10min
flow rate 2 ml/min
injection 10muL, 20 µg/mL each antioxidant
detector UV, 280 nm
Description
Categories Analytical Chromatography, Antioxidants
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC