Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Antiprotozoal Drugs on Ascentis® Phenyl
HPLC Analysis of Antiprotozoal Drugs on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 C
mobile phase [A] 10 mM ammonium acetate (pH 5.0 with acetic acid); [B] acetonitrile; (60:40, A:B)
flow rate 1.0 ml/min
sample 50 µg/mL each in 85:15, water:methanol
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antiprotozoal drugs, Antibiotics, Antimicrobials