HPLC Analysis of Antipyretics/Analgesics/Antifungals on Discovery® RP-AmideC16
HPLC Analysis of Antipyretics/Analgesics/Antifungals on Discovery® RP-AmideC16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 30 °C
mobile phase 0.1% TFA in (25:75) acetonitrile:water
flow rate 1 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Analgesics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC