Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Antitussive/Antihistamine/Antipyretic Mix on Discovery® RP-Amide C16
HPLC Analysis of Antitussive/Antihistamine/Antipyretic Mix on Discovery® RP-Amide C16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. ambient
mobile phase [A] 25 mM potassium phosphate (pH 2.3); [B] acetonitrile
flow rate 2 ml/min
injection 10 µL, each compound 1 µg/mL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antihistamine, Antihistamines
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC