HPLC Analysis of Antiviral Drugs on Ascentis® C8
HPLC Analysis of Antiviral Drugs on Ascentis® C8
Conditions
column Ascentis C8, 15 cm x 2.1 mm I.D., 3 µm particles (581402-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium phosphate, pH 2.5 with phosphoric acid; [B] acetonitrile
gradient 5 to 80% B in 10 min
flow rate 0.2 mL/min
sample as indicated in mobile phase
injection 2 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, antiviral, Antibiotics, Antimicrobials, antibiotics/antimicrobials
Featured Industry Clinical
Pharmaceutical (small molecule)