HPLC Analysis of Anxiolytic Drugs on SUPELCOSIL™ LC-8
HPLC Analysis of Anxiolytic Drugs on SUPELCOSIL™ LC-8
Conditions
column SUPELCOSIL LC-8, 15 cm x 4.6 mm I.D., 5 µm particles (58220-U)
column temp. 30 °C
mobile phase methanol:acetonitrile:0.005 M KH2PO4 and 0.1 M ammonium acetate buffer (pH 6.0 with glacial acetic acid),26.5:16.5:57 (v/v/v)
flow rate 2 mL/min
injection 10 µL
detector UV, 245 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure, Anxiolytics, Sedatives
Featured Industry Pharmaceutical (small molecule)
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC