HPLC Analysis of Anxiolytic Drugs on Ascentis® Phenyl
HPLC Analysis of Anxiolytic Drugs on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 C
mobile phase [A] 10 mM ammonium acetate (pH 5.5 with acetic acid): [B] acetonitrile; (40:60, A:B)
flow rate 1.0 ml/min
sample 50 µg/mL in 90:10, water:methanol
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Anxiolytics, Sedatives