HPLC Analysis of AQC-Alanine Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of AQC-Alanine Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 25 °C
mobile phase [A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (50:50, A:B)
flow rate 0.9 mL/min
sample 2 mg/mL in methanol
injection 0.5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, AQC-Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC