HPLC Analysis of AQC-Amphetamine Enantiomers on Astec® CYCLOBOND® I 2000 DMP
HPLC Analysis of AQC-Amphetamine Enantiomers on Astec® CYCLOBOND® I 2000 DMP
Conditions
column CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 µm particles (20724AST)
column temp. 30 °C
mobile phase [A] 10 mM ammonium acetate, pH 4.1; [B] acetonitrile; (80:20, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Chiral Applications, Drugs of Abuse, Clinical, Forensic & Toxicology, antiparkinsonian, drugs of abuse, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Forensics and Toxicology
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC