HPLC Analysis of Aripiprazole and Dehydro Aripiprazole Using a Titan™ C18 Column
HPLC Analysis of Aripiprazole and Dehydro Aripiprazole Using a Titan™ C18 Column
Conditions
column Titan C18, 10 cm x 3.0 mm I.D., 1.9 µm particles (577160-U)
column temp. 35 C
mobile phase [A] 10 mM ammonium formate; pH 3.0 with formic acid [B] 0.1% formic acid in methanol
gradient 40% B for 0.5 minutes followed by a linear gradient to 95% B for 3.5 minutes, followed by a hold at 95% B for 2 minutes.
flow rate 0.5 ml/min
pressure 6500 psi (445 bar)
sample 50 µg/mL in 10% methanol
injection 2 µl
detector UV, 245 nm
Description
Analysis Note Aripiprazole is marketed as the atypical antipsychotic Abilify® for the treatment of schizophrenia, bipolar disorder and clinical depression. Dehydroapiprazole is the primary metabolite of Aripiprazole.
Categories Analytical Chromatography, Antidepressants
Featured Industry Pharmaceutical (small molecule)
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC, Abilify is a registered trademark of Otsuka Pharmaceutical Co.