Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ascorbic Acid in White Wine: Scaling Separations from Analytical to Semi-Preparative on SeQuant ZIC-HILIC, 4.6 mm to 20 mm I.D. Columns
HPLC Analysis of Ascorbic Acid in White Wine: Scaling Separations from Analytical to Semi-Preparative on SeQuant ZIC-HILIC, 4.6 mm to 20 mm I.D. Columns
Conditions
column SeQuant ZIC-HILIC, SS, 150 x 4.6 mm or 20 mm, 5 μm, 200 Å (1.50497.0001, 1.50455.0001)
mobile phase [A] 0.1% (v/v) formic acid; [B] acetonitrile; (25:75, A:B)
flow rate 20 ml/min
flow rate 1.0 ml/min
pressure 720 psi (5.0 MPa)
injection 250 and 5000 µl of white wine diluted 1:3 with acetonitrile
detector 240 nm
Description
Featured Industry Food and Beverages