Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Asparaginase on Zenix® SEC-150
HPLC Analysis of Asparaginase on Zenix® SEC-150
Conditions
column Zenix SEC-150, 30 cm x 7.8 mm I.D., 3 µm particles (Z777018)
column temp. ambient
mobile phase 150 mM sodium phosphate, pH 7.0
flow rate 0.6 ml/min
pressure 1015 psi (70 bar)
sample asparaginase (2.0 mg/mL in water), RT 10.895 minute, MW 5000-6000, pI 4.85
inj. temp. 5 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies