HPLC Analysis of Asparagine Enantiomers on Astec® CHIROBIOTIC® T (23 °C)
HPLC Analysis of Asparagine Enantiomers on Astec® CHIROBIOTIC® T (23 °C)
Conditions
sample preparation 1 mg/mL in methanol
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 °C
mobile phase [A] water; [B] ethanol; (50:50, A:B)
flow rate 1 mL/min
injection 5 µL
detector UV, 205 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Native Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC