HPLC Analysis of Aspartic Acid Enantiomers on Astec® CHIROBIOTIC® T, Ethanol Mobile Phase
HPLC Analysis of Aspartic Acid Enantiomers on Astec® CHIROBIOTIC® T, Ethanol Mobile Phase
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 °C
mobile phase [A] water; [B] ethanol; [C] formic acid; (20:80:0.02, A:B:C)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 205 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, Native Amino Acids, All Amino Acids
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC