Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Aspirin and Salicylic Acid on Ascentis® Express RP-Amide
HPLC Analysis of Aspirin and Salicylic Acid on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 4.6 mm I.D., 2.7 µm particles (53929-U)
column temp. 30 C
mobile phase [A] water with 0.1% TFA; [B] acetonitrile with 0.1% TFA; (65:35, A:B)
flow rate 1 ml/min
pressure 2200 psi (152 bar)
sample 50 µg/mL in 75:25, water:acetonitrile
injection 5 µl
detector UV, 250 nm
Description
Analysis Note Salicylic acid is the primary metabolite of aspirin, and is likely responsible for its antiinflammatory properties via suppression of cyclooxygenase genes; it also is thought to suppress colonic carcinogenesis. Ascentis Express RP-Amide provides good retention and resolution at low pH by a combination of hydrophobic and H-bonding intereactions.
Categories Analytical Chromatography, Anti-inflammatories, Analgesics