Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Aswagandha Extract on Discovery® C18
HPLC Analysis of Aswagandha Extract on Discovery® C18
Conditions
column Discovery C18, 25 cm x 4.6 mm I.D., 5 µm particles (504971)
column temp. 27 C
mobile phase [A] 1 mM potassium phosphate, monobasic; [B] acetonitrile
gradient 5 to 45% B in 18 min; 45 to 80% B in 7 min; held at 80% B for 3 min
flow rate 1.5 ml/min
pressure 3191 psi (220 bar)
sample 20 µg/mL in water: acetonitrile (86:14)
injection 20 µl
detector UV, 227 nm
Description
Analysis Note This application demonstrates the suitability of Discovery C18 for the efficient separation of withania extract by the USP monograph.
Categories Analytical Chromatography, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC