HPLC Analysis of Barbiturates on Discovery® C18
HPLC Analysis of Barbiturates on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. ambient
mobile phase [A] acetonitrile; [B] 25 mM potassium phosphate buffer, pH 6.9 (25:75, A:B)
flow rate 1.5 mL/min
injection 25 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Drugs of Abuse, drugs of abuse
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC