HPLC Analysis of Barbiturates on Discovery® C18, Unbuffered Mobile Phase
HPLC Analysis of Barbiturates on Discovery® C18, Unbuffered Mobile Phase
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. ambient
mobile phase [A] methanol; [B] water (45:55, A:B)
flow rate 1 mL/min
injection 10 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Pharmaceutical Analysis, drugs of abuse, Anticonvulsant/antiepiletic/antiseizure, Drugs of Abuse, antiepileptic/antiseizure
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC