HPLC Analysis of Barbiturates on Discovery® RP-AmideC16
HPLC Analysis of Barbiturates on Discovery® RP-AmideC16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 30 °C
mobile phase [A] methanol:acetonitrile, 60:40; [B] water; (25:75, A:B)
flow rate 2 mL/min
injection 2 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure, Drugs of Abuse, antiepileptic/antiseizure, drugs of abuse
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC