HPLC Analysis of Barbiturates on Discovery® C8
HPLC Analysis of Barbiturates on Discovery® C8
Conditions
column Discovery C8, 15 cm x 4.6 mm I.D., 5 µm particles (59353-U)
column temp. ambient
mobile phase [A] methanol; [B] water (45:55, A:B)
flow rate 1 ml/min
injection 5 µl
detector UV, 214nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure, Drugs of Abuse
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC