Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Basic Drug Panel on Ascentis® Express C18
HPLC Analysis of Basic Drug Panel on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 4.6 mm I.D., 2.7 µm particles (53827-U)
column temp. ambient
mobile phase [A] water with 0.05M potassium phosphate and 0.1% TEA and 0.6% OSA-Na at pH = 2.9, [B] acetonitrile
flow rate 1.76 ml/min
injection 100 µl
detector UV, 215 nm
Description
Categories Analytical Chromatography, Antihypertensives, Antidepressants, Antipsychotics