Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Basic Peptides (Angiotensin Analogs) on BIOshell™ A160 Peptide C18
HPLC Analysis of Basic Peptides (Angiotensin Analogs) on BIOshell™ A160 Peptide C18
Conditions
column BIOshell A160 Peptide C18, 10 cm x 3 mm I.D., 2.7 µm particles (66908-U)
column temp. 35 C
mobile phase [A] 0.1% (v/v) formic acid, pH 4.0 (titrated with ammonium hydroxide); [B] 50:50, (0.175% formic acid in water, pH 4.0) : acetonitrile
gradient 20 to 50% B in 15 min
flow rate 0.6 ml/min
pressure 3002 psi (207 bar)
sample 0.5 g/L ea. peptide
injection 2.0 µl
detector UV, 215 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC