HPLC Analysis of Benzalkonium Chloride in Commercial Disinfecting Wipes on Ascentis® Express HILIC
HPLC Analysis of Benzalkonium Chloride in Commercial Disinfecting Wipes on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 µm particles (53981-U)
column temp. 35 °C
mobile phase [A] 20 mM ammonium acetate; [B] acetonitrile; (10:90, A:B)
flow rate 1 mL/min
sample Single wipe sheet, extracted with 10 mL methanol
injection 2 µL
detector ESI(+), TIC 150 - 500 m/z
Description
Categories Analytical Chromatography, Surfactants
Featured Industry Cosmetics, Personal Care, and Cleaning Products