Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Benzalkonium Chlorides on Discovery® Cyano
HPLC Analysis of Benzalkonium Chlorides on Discovery® Cyano
Conditions
column Discovery Cyano, 15 cm x 4.6 mm I.D., 5 µm particles (59356-U)
column temp. ambient
mobile phase [A] acetonitrile; [B] acetate buffer, pH 4.5, (60:40, A:B)
flow rate 1 ml/min
sample undefined C8 to C18 Benzalkonium Chlorides
detector UV, 254 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC