HPLC Analysis of Benzene and Deuterated Benzene on Ascentis® Express C18
HPLC Analysis of Benzene and Deuterated Benzene on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm particles (53829-U)
column temp. 50 °C
mobile phase [A] acetonitrile; [B] water (55:45, A:B)
flow rate 1.0 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Petroleum