HPLC Analysis of Benzodiazepines on Ascentis® C8
HPLC Analysis of Benzodiazepines on Ascentis® C8
Conditions
column Ascentis C8, 15 cm x 2.1 mm I.D., 3 µm particles (581402-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate (pH 6.98 unadjusted); [B] acetonitrile; (55:45, A:B)
flow rate 0.2 mL/min
sample 1 µg/mL each in 10 mM ammonium acetate (pH 6.98 unadjusted)
injection 5 µL
detector MS, ESI (+), Selected Ion Recording Mode
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, psychotropics, Anticonvulsant/antiepiletic/antiseizure, antiepileptic/antiseizure, Psychotropics
Featured Industry Clinical
Pharmaceutical (small molecule)