HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl with Acetonitrile Mobile Phase
HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl with Acetonitrile Mobile Phase
Conditions
column Ascentis Express Phenyl-Hexyl, 5 cm x 4.6 mm I.D., 2.7 µm particles (53348-U)
column temp. 35 C
mobile phase [A] water; [B] acetonitrile; (62:38, A:B)
flow rate 1.4 ml/min
pressure 2600 psi (179 bar)
sample 0.1 g/mL of each in 20% methanol
injection 2 µl
detector UV, 250 nm
Description
Analysis Note The Phenyl-Hexyl phase of the Ascentis Express product line displays the best selectivity of this sample; selectivity is markedly affected by choice of organic modifier. The application also demonstrates feasibility of fast LC with Ascentis Express by virtue of low backpressure and high efficiency.
Categories Analytical Chromatography, Pharmaceutical Analysis, Clinical, antipsychotics, antiepileptic/antiseizure, Anticonvulsant/antiepiletic/antiseizure, Antipsychotics
Featured Industry Clinical
Pharmaceutical (small molecule)