HPLC Analysis of Benzoic Acid and Related Compounds on Ascentis® RP-Amide
HPLC Analysis of Benzoic Acid and Related Compounds on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] water with 0.1% TFA; [B] acetonitrile; (70:30, A:B)
flow rate 1 mL/min
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Preservatives
Featured Industry Cosmetics, Personal Care, and Cleaning Products