Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Benzoic Acid and Sorbic Acid on Ascentis® Express RP-Amide ("Green" Mobile Phase Conditions)
HPLC Analysis of Benzoic Acid and Sorbic Acid on Ascentis® Express RP-Amide (
Conditions
column Ascentis Express RP-Amide, 5 cm x 3 mm I.D., 2.7 µm particles (53916-U)
column temp. 55 C
mobile phase [A] 10 mM ammonium phosphate monobasic, pH 2.5 with phosphoric acid; [B] ethanol; (75:25, A:B)
flow rate 0.4 ml/min
pressure 138 bar (2000 psi) column, 248 bar (3600 psi) total
sample 50 µg/mL in mobile phase
injection 1 µl
detector UV, 215 nm
Description
Categories Analytical Chromatography, Organic Acids
Featured Industry Cosmetics, Personal Care, and Cleaning Products