Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Benzoic Acid Derivatives on Ascentis® C18
HPLC Analysis of Benzoic Acid Derivatives on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 30 C
mobile phase [A] 10mM TFA in water; [B] 10 mM TFA in acetonitrile; (70:30, A:B)
flow rate 1.0 ml/min
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Carboxylic acids
Featured Industry Vitamins, Nutraceuticals, and Natural Products